siRNA Details
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014116.1 | NP_001014138.1 | 2, 3 | 412 | ||
XM_006245998.2 | XP_006246060.1 | 2, 3 | 428 | |||
XM_006245999.2 | XP_006246061.1 | 2, 3 | 428 | |||
GenBank mRNA | BC079205.1 | - | 2, 3 | 412 | 7131778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
