siRNA Details
Gene Symbol: RGD1311343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108270.1 | NP_001101740.1 | 2 | 337 | ||
XM_006246018.2 | XP_006246080.1 | 2 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
