siRNA Details
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108272.2 | NP_001101742.1 | 2, 3 | 440 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145962, AK039865, AK136305, BC027089, BC032188, and others.
Gene Ontology Categories:
