siRNA Details
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002022.1 | NP_001002022.1 | 4 | 381 | ||
XM_006246149.1 | XP_006246211.1 | 4 | 375 | |||
XM_008767688.1 | XP_008765910.1 | 3 | 4871 | |||
XM_008767689.1 | XP_008765911.1 | 4 | 283 | |||
GenBank mRNA | BC076397.1 | - | 4 | 381 | 7109965 |

More Information
Other Species Targeted

Gene Ontology Categories:
