siRNA Details
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002022.1 | NP_001002022.1 | 3 | 298 | ||
XM_006246149.1 | XP_006246211.1 | 3 | 292 | |||
XM_008767688.1 | XP_008765910.1 | 2 | 4788 | |||
XM_008767689.1 | XP_008765911.1 | 3 | 200 | |||
GenBank mRNA | BC076397.1 | - | 3 | 298 | 7109965 |
Gene Symbol: LOC317516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773226.1 | XP_008771448.1 | 1 | 219 |
