siRNA Details
Gene Symbol: P4ha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108275.1 | NP_001101745.1 | 12 | 1563 | ||
XM_006246300.1 | XP_006246362.1 | 12 | 1532 | |||
XM_008767691.1 | XP_008765913.1 | 12 | 1560 | |||
GenBank mRNA | FQ228519.1 | - | 12 | 1468 |
