siRNA Details
Gene Symbol: Slc47a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014118.2 | NP_001014140.1 | 2, 3 | 236 | ||
GenBank mRNA | AB248823.1 | - | 2, 3 | 236 | ||
AB248824.1 | - | 2, 3 | 236 | |||
BC088413.1 | - | 256 | 7109897 | |||
FQ217802.1 | - | 250 |

More Information
Other Species Targeted

Gene Ontology Categories:
