siRNA Details
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114606.1 | NP_001108078.1 | 1, 2 | 251 | ||
GenBank mRNA | BC081953.1 | - | 1, 2 | 224 | 7122794 |

More Information
Other Species Targeted

Gene Ontology Categories:
