siRNA Details
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008361.1 | NP_001008362.1 | 7 | 1219 | ||
XM_006246750.2 | XP_006246812.1 | 7 | 633 | |||
GenBank mRNA | BC083670.1 | - | 7 | 1219 | 7135651 | |
FQ216790.1 | - | 7 | 553 | |||
FQ225667.1 | - | 4 | 249 | |||
FQ234995.1 | - | 4 | 249 |
