siRNA Details
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113781.2 | NP_001107253.2 | 6 | 1273 | ||
XM_006246759.2 | XP_006246821.1 | 6 | 1383 | |||
XM_006246760.2 | XP_006246822.1 | 6 | 1431 | |||
GenBank mRNA | BC158692.1 | - | 6 | 1273 | 9022735 |
