siRNA Details
Gene Symbol: RGD1304728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246849.2 | XP_006246911.1 | 18 | 2999 | ||
XM_008767904.1 | XP_008766126.1 | 18 | 2848 | |||
XM_008767905.1 | XP_008766127.1 | 18 | 2764 | |||
XM_008767906.1 | XP_008766128.1 | 18 | 2972 | |||
XM_008767907.1 | XP_008766129.1 | 17 | 2903 | |||
XM_340837.6 | XP_340838.3 | 18 | 2999 |
