siRNA Details
Gene Symbol: Tax1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025419.1 | NP_001020590.1 | 2 | 106 | ||
XM_006246761.2 | XP_006246823.1 | 2 | 313 | |||
GenBank mRNA | BC097976.1 | - | 2 | 106 | 7442643 | |
FQ215653.1 | - | 2 | 125 | |||
FQ220071.1 | - | 2 | 125 | |||
FQ220113.1 | - | 2 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
