siRNA Details
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191824.1 | NP_001178753.1 | 5 | 353 | ||
XM_006246762.2 | XP_006246824.1 | 6 | 559 | |||
XM_006246763.2 | XP_006246825.1 | 6 | 453 | |||
GenBank mRNA | AB236891.1 | - | 6 | 547 |
