siRNA Details
Gene Symbol: Ksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108284.1 | NP_001101754.1 | 8 | 1326 | ||
XM_006246924.2 | XP_006246986.1 | 9 | 1549 | |||
XM_006246926.2 | XP_006246988.1 | 9 | 1547 | |||
XM_006246927.2 | XP_006246989.1 | 9 | 1547 | |||
XM_008768063.1 | XP_008766285.1 | 8 | 1634 | |||
XM_008768064.1 | XP_008766286.1 | 8 | 1633 | |||
XM_008768065.1 | XP_008766287.1 | 8 | 1633 | |||
XM_008768066.1 | XP_008766288.1 | 7 | 1609 | |||
XM_008768067.1 | XP_008766289.1 | 9 | 1548 |
