siRNA Details
Gene Symbol: Ankrd13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768134.1 | XP_008766356.1 | 7 | 954 | ||
XM_008768135.1 | XP_008766357.1 | 7 | 957 | |||
XM_008768136.1 | XP_008766358.1 | 7 | 953 | |||
XM_008768137.1 | XP_008766359.1 | 7 | 953 | |||
XM_008768138.1 | XP_008766360.1 | 7 | 953 |

More Information
Other Species Targeted

