siRNA Details
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 3 | 632 | ||
XM_006247047.2 | XP_006247109.1 | 4 | 714 | |||
XM_006247048.2 | XP_006247110.1 | 4 | 713 | |||
XM_006247049.2 | XP_006247111.1 | 4 | 711 | |||
XM_006247050.2 | XP_006247112.1 | 4 | 710 | |||
XM_006247051.2 | XP_006247113.1 | 4 | 712 | |||
XM_006247052.2 | XP_006247114.1 | 4 | 712 | |||
XM_006247053.2 | XP_006247115.1 | 4 | 709 | |||
XM_006247054.2 | XP_006247116.1 | 4 | 708 | |||
XM_008768070.1 | XP_008766292.1 | 4 | 688 | |||
XM_008768071.1 | XP_008766293.1 | 4 | 658 | |||
GenBank mRNA | AY633742.1 | - | 3 | 632 |

More Information
Other Species Targeted

Gene Ontology Categories:
