siRNA Details
Gene Symbol: Ints2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100630.2 | NP_001094100.2 | 23 | 3542 | ||
XM_008768072.1 | XP_008766294.1 | 23 | 3609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
