siRNA Details
Gene Symbol: Ypel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108286.1 | NP_001101756.1 | 2 | 366 | ||
XM_006247097.1 | XP_006247159.1 | 2 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
