siRNA Details
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108287.1 | NP_001101757.1 | 7 | 889 | ||
XM_008768073.1 | XP_008766295.1 | 7 | 1046 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108287.1 | NP_001101757.1 | 7 | 889 | ||
XM_008768073.1 | XP_008766295.1 | 7 | 1046 |