siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108291.1 | NP_001101761.1 | 11 | 1576 | ||
XM_008768078.1 | XP_008766300.1 | 10 | 1672 | |||
GenBank mRNA | FQ212347.1 | - | 10 | 1652 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026313, BC009092.
Gene Ontology Categories:
