siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108291.1 | NP_001101761.1 | 4 | 391 | ||
XM_006247155.2 | XP_006247217.1 | 4 | 383 | |||
XM_006247156.2 | XP_006247218.1 | 4 | 382 | |||
XM_008768078.1 | XP_008766300.1 | 4 | 382 | |||
GenBank mRNA | FQ212347.1 | - | 4 | 364 | ||
FQ231407.1 | - | 2 | 41 | |||
FQ231682.1 | - | 4 | 364 | |||
FQ235074.1 | - | 4 | 357 |
