siRNA Details
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247233.2 | XP_006247295.1 | 1, 2 | 230 | ||
XM_006247235.2 | XP_006247297.1 | 1, 2 | 229 |

More Information
Other Species Targeted

Gene Ontology Categories:
