siRNA Details
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247236.2 | XP_006247298.1 | 2 | 831 | ||
XM_006247237.2 | XP_006247299.1 | 2 | 823 | |||
GenBank mRNA | BC169095.1 | - | 225 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008686, NM_001130450, BC047283, AK154167, AF071084, and others.
Gene Ontology Categories:
