siRNA Details
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247236.2 | XP_006247298.1 | 3 | 1182 | ||
XM_006247237.2 | XP_006247299.1 | 3 | 1174 | |||
XM_006247238.2 | XP_006247300.1 | 2 | 344 | |||
XM_006247239.2 | XP_006247301.1 | 2 | 97 | |||
GenBank mRNA | BC169095.1 | - | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
