siRNA Details
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012142.1 | NP_001012142.1 | 2 | 135 | ||
XM_006247342.2 | XP_006247404.1 | 2 | 364 | |||
XM_008768079.1 | XP_008766301.1 | 2 | 1648 | |||
GenBank mRNA | BC078939.1 | - | 2 | 135 | 7108372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
