siRNA Details
Gene Symbol: Krt19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY464140.1 | - | 4 | 877 | ||
BC088424.1 | - | 4 | 873 | |||
BC126075.1 | - | 4 | 868 | 7442082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
