siRNA Details
Gene Symbol: Fkbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014120.1 | NP_001014142.1 | 3 | 565 | ||
GenBank mRNA | BC085854.1 | - | 3 | 565 | 7190051 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fkbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014120.1 | NP_001014142.1 | 3 | 565 | ||
GenBank mRNA | BC085854.1 | - | 3 | 565 | 7190051 |