siRNA Details
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108309.1 | NP_001101779.1 | 9, 10 | 1404 | ||
GenBank mRNA | FQ212255.1 | - | 9, 10 | 1314 |

More Information
Other Species Targeted

Gene Ontology Categories:
