siRNA Details
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007005.1 | NP_001007006.1 | 2 | 122 | ||
XM_006247897.2 | XP_006247959.1 | 2 | 256 | |||
XM_006247898.2 | XP_006247960.1 | 2 | 205 | |||
GenBank mRNA | BC083817.1 | - | 2 | 122 | 7110945 |
