siRNA Details
Gene Symbol: LOC102550455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247957.2 | XP_006248019.1 | 1 | 488 |
Gene Symbol: Narf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039207.2 | NP_001034296.1 | 8 | 881 | ||
XM_006247899.2 | XP_006247961.1 | 8 | 914 | |||
GenBank mRNA | BC110050.1 | - | 8 | 881 | 7454395 |

More Information
Other Species Targeted

Gene Ontology Categories:
