siRNA Details
Gene Symbol: Narf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039207.2 | NP_001034296.1 | 10 | 1148 | ||
XM_006247899.2 | XP_006247961.1 | 10 | 1181 | |||
GenBank mRNA | BC110050.1 | - | 10 | 1148 | 7454395 |
Gene Symbol: LOC102550455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247957.2 | XP_006248019.1 | 3 | 755 |
