siRNA Details
Gene Symbol: Gtpbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025015.1 | NP_001020186.1 | 5, 6 | 794 | ||
XM_006248340.2 | XP_006248402.1 | 4, 5 | 720 | |||
GenBank mRNA | BC091236.1 | - | 5, 6 | 794 | 7322967 | |
FQ216550.1 | - | 5, 6 | 793 |
