siRNA Details
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751052.3 | XP_003751100.1 | 17 | 3075 | ||
XM_006248353.2 | XP_006248415.1 | 17 | 3081 | |||
XM_006248354.2 | XP_006248416.1 | 17 | 3078 | |||
XM_006248355.2 | XP_006248417.1 | 17 | 3078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
