siRNA Details
Gene Symbol: Ostn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207612.2 | NP_997495.1 | 2, 3 | 323 | ||
XM_008768801.1 | XP_008767023.1 | 3, 4 | 515 | |||
XM_008768803.1 | XP_008767025.1 | 4, 5 | 665 | |||
GenBank mRNA | AY398682.1 | - | 325 | |||
AY573934.1 | - | 2, 3 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
