siRNA Details
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108323.1 | NP_001101793.1 | 9 | 903 | ||
XM_006248628.2 | XP_006248690.1 | 9 | 1028 | |||
XM_006248629.2 | XP_006248691.1 | 9 | 898 | |||
XM_006248630.2 | XP_006248692.1 | 9 | 923 | |||
XM_006248633.2 | XP_006248695.1 | 9 | 1027 | |||
XM_006248634.2 | XP_006248696.1 | 8 | 924 | |||
XM_008768888.1 | XP_008767110.1 | 9 | 901 | |||
GenBank mRNA | FQ213583.1 | - | 8 | 930 |
