siRNA Details
Gene Symbol: Lztr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108326.2 | NP_001101796.1 | 16 | 1830 | ||
XM_006248707.2 | XP_006248769.1 | 16 | 2143 | |||
GenBank mRNA | BC169071.1 | - | 16 | 1830 | 7938530 | |
FQ211947.1 | - | 16 | 1885 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
