siRNA Details
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248842.2 | XP_006248904.1 | 6 | 1239 | ||
XM_006248844.2 | XP_006248906.1 | 6 | 1239 | |||
XM_006248845.2 | XP_006248907.1 | 6 | 1242 | |||
XM_346914.7 | XP_346915.4 | 5 | 1076 |

More Information
Other Species Targeted

Gene Ontology Categories:
