siRNA Details
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248842.2 | XP_006248904.1 | 7 | 1572 | ||
XM_006248844.2 | XP_006248906.1 | 7 | 1572 | |||
XM_006248845.2 | XP_006248907.1 | 7 | 1575 | |||
XM_346914.7 | XP_346915.4 | 6 | 1409 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028291, BC082547, CB722719, AK046685, CF743030.
Gene Ontology Categories:
