siRNA Details
Gene Symbol: Zfp655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008362.1 | NP_001008363.1 | 4 | 1163 | ||
XM_006248895.2 | XP_006248957.1 | 4 | 1257 | |||
XM_008768977.1 | XP_008767199.1 | 4 | 1529 | |||
XM_008768978.1 | XP_008767200.1 | 4 | 1618 | |||
GenBank mRNA | BC085937.1 | - | 4 | 1163 | 7121505 |
Gene Symbol: LOC103690102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769057.1 | XP_008767279.1 | 3 | 1072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
