siRNA Details
Gene Symbol: Ap1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108331.2 | NP_001101801.2 | 2 | 167 | ||
XM_006249179.2 | XP_006249241.1 | 2 | 196 | |||
GenBank mRNA | BC169070.1 | - | 2 | 111 | 5598714 | |
FQ230081.1 | - | 1 | 31 |
