siRNA Details
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037788.1 | NP_001032877.1 | 2 | 100 | ||
XM_006249180.2 | XP_006249242.1 | 2 | 320 | |||
XM_006249181.2 | XP_006249243.1 | 2 | 282 | |||
XM_008769143.1 | XP_008767365.1 | 2 | 279 | |||
XM_008769144.1 | XP_008767366.1 | 1 | 32 | |||
XM_008769146.1 | XP_008767368.1 | 2 | 320 | |||
XM_008769148.1 | XP_008767370.1 | 2 | 320 | |||
GenBank mRNA | BC098678.1 | - | 2 | 100 | 7382696 |
