siRNA Details
Gene Symbol: Sumf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025125.2 | NP_001020296.2 | 7, 8 | 701 | ||
GenBank mRNA | BC091176.1 | - | 7, 8 | 707 | 7322044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
