siRNA Details
Gene Symbol: Snrnp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014127.1 | NP_001014149.1 | 2 | 615 | ||
XM_006249325.2 | XP_006249387.1 | 2 | 663 | |||
GenBank mRNA | BC086435.1 | - | 2 | 615 | 7302221 |
