siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134958.1 | NP_001128430.1 | 7 | 1486 | ||
XM_006249782.2 | XP_006249844.1 | 7 | 1573 | |||
XM_006249783.2 | XP_006249845.1 | 7 | 1457 | |||
XM_006249784.2 | XP_006249846.1 | 7 | 1574 | |||
XM_006249785.2 | XP_006249847.1 | 6 | 2522 | |||
XM_008769479.1 | XP_008767701.1 | 7 | 1568 | |||
XM_008769480.1 | XP_008767702.1 | 4 | 490 | |||
XM_008769481.1 | XP_008767703.1 | 4 | 419 | |||
GenBank mRNA | BC168690.1 | - | 7 | 1486 | 7934193 |
