siRNA Details
Gene Symbol: Snrpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271237.1 | NP_001258166.1 | 4 | 181 | ||
GenBank mRNA | FQ211122.1 | - | 4 | 181 |

More Information
Other Species Targeted

Gene Ontology Categories:
