siRNA Details
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109667.1 | NP_001103137.1 | 3, 4 | 436 | ||
GenBank mRNA | BC128780.1 | - | 3, 4 | 436 | 7315719 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145513, AK049119, BC058250, BC002098, AK049784, and others.
Gene Ontology Categories:
