siRNA Details
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109667.1 | NP_001103137.1 | 4, 5 | 557 | ||
GenBank mRNA | BC128780.1 | - | 4, 5 | 557 | 7315719 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109667.1 | NP_001103137.1 | 4, 5 | 557 | ||
GenBank mRNA | BC128780.1 | - | 4, 5 | 557 | 7315719 |