siRNA Details
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012150.1 | NP_001012150.1 | 3 | 362 | ||
XM_006250393.2 | XP_006250455.1 | 3 | 596 | |||
XM_006250400.2 | XP_006250462.1 | 3 | 596 | |||
XM_006250401.2 | XP_006250463.1 | 3 | 596 | |||
XM_008769859.1 | XP_008768081.1 | 3 | 492 | |||
XM_008769860.1 | XP_008768082.1 | 3 | 1122 | |||
XM_008769861.1 | XP_008768083.1 | 3 | 492 | |||
GenBank mRNA | BC083927.1 | - | 3 | 362 | 7135538 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001083120, NM_001083121, NM_008680, NM_010135, BC062927, and others.
Gene Ontology Categories:
