siRNA Details
Gene Symbol: Taf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037204.1 | NP_001032281.1 | 7 | 966 | ||
XM_008769862.1 | XP_008768084.1 | 7 | 988 | |||
GenBank mRNA | BC107466.1 | - | 7 | 966 | 7460374 |
