siRNA Details
Gene Symbol: Esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203336.2 | NP_976081.1 | 5 | 920 | ||
XM_006250443.2 | XP_006250505.1 | 4 | 1573 | |||
GenBank mRNA | AB126962.1 | - | 4 | 804 | ||
AB126963.1 | - | 5 | 1032 | |||
AY341057.1 | - | 4 | 683 |
